Specifications
- Antibody type:Primary
- Antigen name:BCL2-like 1
- Antigen symbol:BCL2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:BCLX|BCL2L1|Apoptosis regulator Bcl-X|BLC2L|BCL2L|Bcl-2-like 1 protein
- Amino acid number:120 to 130
- UniProtKB:Q07817
- Sequence:YQSFEQDTFVE
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Bcl-XL (120-130aa YQSFEQDTFVE), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Bcl-2-like protein 1(BCL2L1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Bcl-xS is the second mRNA species of Bcl-x protein, which encodes a protein that inhibits the ability of bcl-2 to enhance the survival of growth factor-deprived cells. In vivo, bcl-xS mRNA is expressed at high levels in cells that undergo a high rate of turnover, such as developing lymphocytes. Apoptosis was associated with increased protein expression of Bcl-X/S, an early marker of apoptosis, and release of cytochrome c into the cytosol of infected HB cells.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Bcl-XL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat