Anti-OPCML Rabbit Polyclonal Antibody
Catalog # BSBTPA1131
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Opioid binding Protein/Cell Adhesion molecule-like
- Antigen symbol:OPCML
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:IGLON1|OBCAM|OPCM
- Amino acid number:294 - 312
- Sequence:YTCVATNKLGNTNASITLY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human OPCML (294-312aa YTCVATNKLGNTNASITLY), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Opioid-binding protein/cell adhesion molecule (OPCML) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: OPCML
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat