Anti-NSE Rabbit Polyclonal Antibody
Catalog # BSBTPA1061
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Enolase 2
- Antigen symbol:ENO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Gamma-enolase|Enolase 2|ENO2|Neural enolase|NSE|2-phospho-D-glycerate hydro-lyase|Neuron-specific enolase
- Amino acid number:420 - 434
- Sequence:EARFAGHNFRNPSVL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human NSE (420-434aa EARFAGHNFRNPSVL), identical to the related mouse and rat sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Gamma-enolase (ENO2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: NSE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat