Anti-NGF Rabbit Polyclonal Antibody
100 µg
Catalog # BSBTPA1056
Supplier: Boster Bio
Jump to:
Product Details & Documents
Rabbit IgG polyclonal antibody for Beta-nerve growth factor (NGF) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Epitope:N-Terminal
- Form:Lyophilised
- Amino acid number:162 - 173
- Storage temperature:–20 °C
- Antigen name:Nerve Growth Factor
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human NGF (162-173aa EVNINNSVFKQY), identical to the related rat sequence, and different from the related mouse sequence by one amino acid
- Purification:purified by immunogen affinity purification
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Sequence:EVNINNSVFKQY
- Cross adsorption:No
- Shipping temperature:Ice
- Cat. No.:BSBTPA1056
- Reactivity:Human|Rat|Mouse
- Antigen synonyms:HSAN5|Beta-NGF|NGFB
- Conjugation:Unconjugated
- Antigen symbol:NGF



