Anti-Talin Mouse Monoclonal Antibody [clone: Tal-33]
Catalog # BSBTMA1092
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Talin 1
- Antigen symbol:TLN1
- Clonality:Monoclonal
- Clone:Tal-33
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:TLN|TLN1|ILWEQ|talin 1|KIAA1027
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Purified chicken gizzard talin.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Talin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine