Order Entry
Export
ContactUsLinkComponent
 
Anti-Spectrin alpha/beta Mouse Monoclonal Antibody [clone: Spe 1/2]
  BSBTMA1090
Anti-Spectrin alpha/beta Mouse Monoclonal Antibody [clone: Spe 1/2]
  BSBTMA1090

 

  • Antibody type:
    Primary
  • Antigen name:
    Spectrin alpha/beta
  • Antigen symbol:
    SPTA1/SPTB
  • Clonality:
    Monoclonal
  • Clone:
    Spe 1/2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Human erythrocyte spectrin.
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: Spectrin alpha/beta
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human