Anti-MAP Kinase, Activated Mouse Monoclonal Antibody [clone: IL-13]
BSBTMA1055
- Antibody type:Primary
- Antigen name:MAP Kinase, Activated
- Clonality:Monoclonal
- Clone:IL-13
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Yeast,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Modification:phospho-Glu, phospho-Val
- Sequence:HTGFLTpEYpVAT
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide containing 11 amino acids, HTGFLTpEYpVAT, corresponding to the phosphorylated form of ERK-activation loop conjugated to KLH.
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: MAP Kinase, Activated
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Yeast