Order Entry
Export
ContactUsLinkComponent
Anti-HSP60 Mouse Monoclonal Antibody [clone: SJ-60]
Anti-HSP60 Mouse Monoclonal Antibody [clone: SJ-60]
Catalog # BSBTMA1049
Supplier:  Boster Bio
Anti-HSP60 Mouse Monoclonal Antibody [clone: SJ-60]
Catalog # BSBTMA1049
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat Shock Protein 60
  • Antigen symbol:
    HSP60
  • Clonality:
    Monoclonal
  • Clone:
    SJ-60
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Chicken,
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human heat shock protein 60 (HSP60)
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Chicken