Anti-Gelsolin Mouse Monoclonal Antibody [clone: GEL-42]
Catalog # BSBTMA1044
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Gelsolin
- Antigen symbol:GSN
- Clonality:Monoclonal
- Clone:GEL-42
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:actin-depolymerizing factor|DKFZp313L0718|AGEL|RP11-477J21.1|brevin|ADF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human plasma gelsolin
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for Gelsolin, gelsolin (GSN) detection. Tested with WB, IHC-P, IHC-F in Human; rabbit; bovine. No cross reactivity with otherproteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Gelsolin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit