Anti-Cyclin D2 Mouse Monoclonal Antibody [clone: CY-D2]
Catalog # BSBTMA1034
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cyclin D2
- Antigen symbol:CCND2
- Clonality:Monoclonal
- Clone:CY-D2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:G1/S-specific cyclin-D2|MPPH3|G1/S-specific cyclin D2|MGC102758|KIAK0002
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human cyclin D2 protein.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cyclin D2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse