Anti-Collagen, Type III Mouse Monoclonal Antibody [clone: Col-29]
Catalog # BSBTMA1029
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Collagen, Type III
- Antigen symbol:Collagen, Type III
- Clonality:Monoclonal
- Clone:Col-29
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human collagen type III
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for Collagen, Type III, collagen, type III, alpha 1 (COL3A1) detection. Tested with WB, IHC-F in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Collagen, Type III
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat