Order Entry
Export
ContactUsLinkComponent
Anti-Calponin Mouse Monoclonal Antibody [clone: CP-19]
Anti-Calponin Mouse Monoclonal Antibody [clone: CP-19]
Catalog # BSBTMA1011
Supplier:  Boster Bio
Anti-Calponin Mouse Monoclonal Antibody [clone: CP-19]
Catalog # BSBTMA1011
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calponin
  • Antigen symbol:
    CNN
  • Clonality:
    Monoclonal
  • Clone:
    CP-19
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Turkey,
    Chicken,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    CNN3|Calponin 1 basic smooth muscle|CNN2|CNN1
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Turkey gizzard calponin
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Calponin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken, Turkey