Order Entry
Export
ContactUsLinkComponent
 
Anti-Actin Mouse Monoclonal Antibody [clone: AC-40]
  BSBTMA1000
undefined
Anti-Actin Mouse Monoclonal Antibody [clone: AC-40]
  BSBTMA1000

 

  • Antibody type:
    Primary
  • Antigen name:
    Actin Alpha 1, Skeletal Muscle
  • Antigen symbol:
    ACTA1
  • Clonality:
    Monoclonal
  • Clone:
    AC-40
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic actin C-terminal peptideSer-Gly-Pro-Ser-Ile-Val-His-Arg-Lys-Cys-Phe,attached to a Multiple Antigen Peptide (MAP) backbone
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Mouse IgG monoclonal antibody for Actin detection. Tested with WB, IHC-F, ICC in Human;mouse;rat;chicken. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Actin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Chicken