Order Entry
Export
ContactUsLinkComponent
 
Anti-eIF5A2 + eIF5A Rabbit Monoclonal Antibody [clone: EPR7412-50]
 :  Abcam
New Product
 
undefined
Anti-eIF5A2 + eIF5A Rabbit Monoclonal Antibody [clone: EPR7412-50]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB150439-1ML
  • ABCAAB150439-100
  • Antigen symbol:
  • EIF5A2
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice