Order Entry
Export
ContactUsLinkComponent
Anti-Annexin-11/ANXA11 Rabbit Polyclonal Antibody
Anti-Annexin-11/ANXA11 Rabbit Polyclonal Antibody
Catalog # ABCAAB137424-50
Supplier:  Abcam
undefined
Anti-Annexin-11/ANXA11 Rabbit Polyclonal Antibody
Catalog # ABCAAB137424-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Annexin A11
  • Antigen symbol:
    ANXA11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ANXA11 (annexin A11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human