Order Entry
Export
ContactUsLinkComponent
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3]
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3]
Catalog # ABCAAB134375-100
Supplier:  Abcam
undefined
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3]
Catalog # ABCAAB134375-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD24 molecule
  • Antigen symbol:
    CD24
  • Clonality:
    Monoclonal
  • Clone:
    SN3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SN3] to CD24.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: SN3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human