Order Entry
Export
ContactUsLinkComponent
 
Anti-BCKDHA Rabbit Polyclonal Antibody
  ABCAAB126173-50
 :  Abcam
undefined
Anti-BCKDHA Rabbit Polyclonal Antibody
  ABCAAB126173-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    branched chain keto acid dehydrogenase E1, alpha polypeptide
  • Antigen symbol:
    BCKDHA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Rabbit polyclonal to BCKDHA.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BCKDHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human