Order Entry
Export
ContactUsLinkComponent
Anti-TMEM61 Rabbit Polyclonal Antibody
Anti-TMEM61 Rabbit Polyclonal Antibody
Catalog # ABCAAB122117-100
Supplier:  Abcam
undefined
Anti-TMEM61 Rabbit Polyclonal Antibody
Catalog # ABCAAB122117-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    transmembrane Protein 61
  • Antigen symbol:
    TMEM61
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal to TMEM61.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TMEM61
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human