Order Entry
Export
ContactUsLinkComponent
 
Anti-TPD52L1 Rabbit Polyclonal Antibody
  ABCAAB121962-100
 :  Abcam
undefined
Anti-TPD52L1 Rabbit Polyclonal Antibody
  ABCAAB121962-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Protein D52-like 1
  • Antigen symbol:
    TPD52L1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal to TPD52L1.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TPD52L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human