Anti-Keratin 36 Rabbit Polyclonal Antibody
ABCAAB111599-50
: Abcam
- Antibody type:Primary
- Antigen name:keratin 36
- Antigen symbol:KRT36
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KRT36 (keratin 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human