Anti-CD163 Mouse Monoclonal Antibody [clone: GHI/61]
Catalog # ABCAAB111250-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:CD163 molecule
- Antigen symbol:CD163
- Clonality:Monoclonal
- Clone:GHI/61
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CD163 (CD163 molecule)
Clonality: monoclonal
Clone: GHI/61
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: