Anti-Cytokeratin 20 Rabbit Monoclonal Antibody [clone: EPR1621(2)]
Supplier: Abcam
New Product
Certificates
About this item
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: monoclonal
Clone: EPR1621
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- ABCAAB109111-100
- ABCAAB109111-1ML
- Antigen symbol:
- KRT20
- Antigen name:
- keratin 20
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- EPR1621
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- ImmunoFluorescence:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice