Order Entry
Export
ContactUsLinkComponent
Anti-G9P Mouse Monoclonal Antibody [clone: mAb 33] (PE (Phycoerythrin))
Anti-G9P Mouse Monoclonal Antibody [clone: mAb 33] (PE (Phycoerythrin))
Catalog # ABCAAB106173-50
Supplier:  Abcam
undefined
Anti-G9P Mouse Monoclonal Antibody [clone: mAb 33] (PE (Phycoerythrin))
Catalog # ABCAAB106173-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4
  • Antigen symbol:
    KIR2DL4
  • Clonality:
    Monoclonal
  • Clone:
    mAb 33
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    0780
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 Tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

Specifications

About this item

PE Mouse monoclonal [mAb 33] to G9P, prediluted.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: G9P
Clonality: Monoclonal
Clone: mAb 33
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human