Anti-IgG2 Mouse Monoclonal Antibody [clone: 31-7-4]
Catalog # ABCAAB99777-250
Supplier: Abcam
Specifications
- Antibody type:Secondary
- Antigen name:IgG2
- Antigen symbol:IgG2
- Clonality:Monoclonal
- Clone:31-7-4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pack type:Vial
- Pk:250 µG
Specifications
About this item
Mouse monoclonal [31-7-4] Anti-Human IgG2 Fc.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Secondary
Antigen: IgG2
Clonality: Monoclonal
Clone: 31-7-4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human