Order Entry
Export
ContactUsLinkComponent
Anti-IgG Mouse Monoclonal Antibody [clone: JDC-10]
Anti-IgG Mouse Monoclonal Antibody [clone: JDC-10]
  ABCAAB99757-250
 :  Abcam
undefined
Anti-IgG Mouse Monoclonal Antibody [clone: JDC-10]
  ABCAAB99757-250
 :  Abcam

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    1370
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pack type:
    Vial
  • Pk:
    250 µG

 

 

Mouse monoclonal [JDC-10] Anti-Human IgG Fc.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: JDC-10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human