Anti-IgD Rat Monoclonal Antibody [clone: 11-26] (FITC)
ABCAAB99566-250
: Abcam
- Antibody type:Secondary
- Antigen symbol:IgD
- Clonality:Monoclonal
- Clone:11-26
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Seq:1370
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pack type:Vial
- Pk:250 µG
Rat monoclonal [11-26] Anti-Mouse IgD delta chain (FITC).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Secondary
Antigen: IgD
Clonality: Monoclonal
Clone: 11-26
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse