Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-CSDE1/NRU Rabbit Polyclonal Antibody

Anti-CSDE1/NRU Rabbit Polyclonal Antibody

50 µl, Vial
Catalog # ABCAAB96124-50
Supplier:   Abcam
Anti-CSDE1/NRU Rabbit Polyclonal Antibody
Catalog # ABCAAB96124-50
Supplier:  Abcam
Anti-CSDE1/NRU Rabbit Polyclonal Antibody
Anti-CSDE1/NRU Rabbit Polyclonal Antibody

50 µl, Vial

Price
569,00 €
Log in for availability
1 * 50 µl
Product Details & Documents

Rabbit Polyclonal CSDE1/NRU antibody. Suitable for WB, IHC-P, ICC/IF and reacts with Human samples. Cited in 3 publications. Immunogen corresponding to Recombinant Fragment Protein within Human CSDE1 aa 150-400.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CSDE1/NRU
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications
  • Seq:
    1202
  • Size:
    50 µl
  • Pack type:
    Vial
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Immunogen Affinity purification
  • Antibody type:
    Primary
  • Form:
    Liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Upon delivery aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
  • Host:
    Rabbit
  • Molecular weight:
    88885 Da
  • Shipping temperature:
    4 °C
  • Antigen name:
    D1S155E
  • Cat. No.:
    ABCAAB96124-50
  • Reactivity:
    Human
  • Antigen synonyms:
    KIAA0885|NRU|UNR|CSDE1|Cold shock domain-containing protein E1|N-ras upstream gene protein|Protein UNR
  • Immunogen:
    Recombinant Fragment Protein within Human CSDE1 aa 150-400. The exact immunogen used to generate this antibody is proprietary information.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Tested applications:
    ICC/IF, IHC-P
  • Antigen symbol:
    CSDE1/NRU
  • Storage buffer:
    Glycerol, Tris, Glycine