Anti-Cardiac Troponin I Mouse Monoclonal Antibody [clone: 19C7]
Catalog # ABCAAB92408-500
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:troponin I type 3 (cardiac)
- Antigen symbol:TNNI3
- Clonality:Monoclonal
- Clone:19C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Seq:1418
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pack type:Vial
- Pk:500 µG
Specifications
About this item
Mouse monoclonal [19C7] to Cardiac Troponin I.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cardiac Troponin I
Clonality: Monoclonal
Clone: 19C7
Conjugation:
Epitope: Recognises an epitope included within aa 41-49
Host: Mouse
Isotype: IgG2b
Reactivity: Human