Order Entry
Export
ContactUsLinkComponent
 
Anti-CACNA1C Mouse Monoclonal Antibody [clone: S57]
  ABCAAB84814-100
 :  Abcam
undefined
Anti-CACNA1C Mouse Monoclonal Antibody [clone: S57]
  ABCAAB84814-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen symbol:
    DHPR alpha 1
  • Clonality:
    Monoclonal
  • Clone:
    S57-46
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [S57] to CACNA1C.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CACNA1C
Clonality: Monoclonal
Clone: S57
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Recombinant full length protein