Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 19 Rabbit Monoclonal Antibody [clone: EPR1579Y]
Anti-Cytokeratin 19 Rabbit Monoclonal Antibody [clone: EPR1579Y]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Cytokeratin 19 Rabbit Monoclonal Antibody [clone: EPR1579Y]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB76539-1ML
  • ABCAAB76539-100
  • Antigen symbol:
  • KRT19
  • Antigen name:
  • keratin 19
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR1579Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications