Order Entry
Export
ContactUsLinkComponent
Anti-HIV1 p17 Mouse Monoclonal Antibody [clone: 17-1]
Anti-HIV1 p17 Mouse Monoclonal Antibody [clone: 17-1]
Catalog # ABCAAB66641-100
Supplier:  Abcam
undefined
Anti-HIV1 p17 Mouse Monoclonal Antibody [clone: 17-1]
Catalog # ABCAAB66641-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HIV1 p17
  • Clonality:
    Monoclonal
  • Clone:
    17-1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal [17-1] to HIV1 p17.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV1 p17
Clonality: Monoclonal
Clone: 17-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: HIV