Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 18 (phospho S52) Rabbit Polyclonal Antibody
Anti-Cytokeratin 18 (phospho S52) Rabbit Polyclonal Antibody
  ABCAAB63393-100
 :  Abcam
undefined
Anti-Cytokeratin 18 (phospho S52) Rabbit Polyclonal Antibody
  ABCAAB63393-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    keratin 18
  • Antigen symbol:
    KRT18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal to Cytokeratin 18 (phospho S52).

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human