Order Entry
Export
ContactUsLinkComponent
Anti-IL-10 Mouse Monoclonal Antibody [clone: B-S10]
Anti-IL-10 Mouse Monoclonal Antibody [clone: B-S10]
  ABCAAB47222-100
 :  Abcam
undefined
Anti-IL-10 Mouse Monoclonal Antibody [clone: B-S10]
  ABCAAB47222-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 10
  • Antigen symbol:
    IL10
  • Clonality:
    Monoclonal
  • Clone:
    B-S10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [B-S10] to IL-10 - BSA and Azide free.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: B-S10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human