Order Entry
Export
ContactUsLinkComponent
 
Anti-Cytokeratin 18 Rabbit Monoclonal Antibody [clone: E431-1]
 :  Abcam
New Product
 
undefined
Anti-Cytokeratin 18 Rabbit Monoclonal Antibody [clone: E431-1]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB32118-100
  • ABCAAB32118-1ML
  • Antigen symbol:
  • KRT18
  • Antigen name:
  • keratin 18
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • E431-1
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice