Specifications
- Antibody type:Primary
- Antigen name:Human Papilloma Virus
- Antigen symbol:HPV16 E1/E4
- Clonality:Monoclonal
- Clone:TVG 403
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Specifications
About this item
Mouse monoclonal [TVG 403] to HPV16 E1+E4.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HPV16 E1+E4
Clonality: Monoclonal
Clone: TVG 403
Conjugation:
Epitope: E1/E4 protein is aa 36-41 of human papilloma virus type 16.
Host: Mouse
Isotype: IgG1
Reactivity: Human papilloma virus (HPV)-16