Order Entry
Export
ContactUsLinkComponent
Anti-JAM-C Rat Monoclonal Antibody [clone: 19 H36]
Anti-JAM-C Rat Monoclonal Antibody [clone: 19 H36]
Catalog # ABCAAB16889-50
Supplier:  Abcam
undefined
Anti-JAM-C Rat Monoclonal Antibody [clone: 19 H36]
Catalog # ABCAAB16889-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Junctional Adhesion Molecule C
  • Antigen symbol:
    JAMC
  • Clonality:
    Monoclonal
  • Clone:
    19 H36
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Rat monoclonal [19 H36] to JAM-C.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: JAM-C
Clonality: Monoclonal
Clone: 19 H36
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse;Human