Order Entry
Export
ContactUsLinkComponent
Anti-EBNA 3C Sheep Polyclonal Antibody
Anti-EBNA 3C Sheep Polyclonal Antibody
Catalog # ABCAAB16128-250
Supplier:  Abcam
Anti-EBNA 3C Sheep Polyclonal Antibody
Catalog # ABCAAB16128-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EBNA 3C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Seq:
    1370
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 μl
  • Pack type:
    Vial
  • Pk:
    250 µl

Specifications

About this item

Sheep polyclonal to EBNA 3C.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EBNA 3C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: