Anti-Cytokeratin 14 Rabbit Polyclonal Antibody
Supplier: Abcam
Certificates
About this item
Rabbit polyclonal to Cytokeratin 14.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cytokeratin 14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCAAB15461-1
- ABCAAB15461-100
- ABCAAB15461-500
- Antigen symbol:
- KRT14
- Antigen name:
- keratin 14
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice