Order Entry
Export
ContactUsLinkComponent
Anti-hCG Holo c2 Mouse Monoclonal Antibody [clone: INN-hCG-53]
Anti-hCG Holo c2 Mouse Monoclonal Antibody [clone: INN-hCG-53]
Catalog # ABCAAB11392-250
Supplier:  Abcam
undefined
Anti-hCG Holo c2 Mouse Monoclonal Antibody [clone: INN-hCG-53]
Catalog # ABCAAB11392-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HCG Holo c2 Epitope
  • Antigen symbol:
    HCG Holo c2
  • Clonality:
    Monoclonal
  • Clone:
    INN-hCG-53
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pack type:
    Vial
  • Pk:
    250 µG

Specifications

About this item

Mouse monoclonal [INN-hCG-53] to hCG Holo c2.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: hCG Holo c2
Clonality: monoclonal
Clone: INN-hCG-53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human