Specifications
- Antibody type:Primary
- Antigen name:Follicle Stimulating Hormone, beta
- Antigen symbol:FSHB
- Clonality:Monoclonal
- Clone:INN-hFSH-60
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Seq:1370
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pack type:Vial
- Pk:250 µG
Specifications
About this item
Mouse monoclonal [INN-hFSH-60] to FSH beta.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FSH beta
Clonality: Monoclonal
Clone: INN-hFSH-60
Conjugation:
Epitope: Directed against the Beta 2 epitope of the Beta subunit of human FSH beta
Host: Mouse
Isotype: IgG1
Reactivity: Human