Order Entry
Export
ContactUsLinkComponent
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ABCAAB9509-250
Supplier:  Abcam
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ABCAAB9509-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
  • Antigen symbol:
    ITGB3
  • Clonality:
    Monoclonal
  • Clone:
    Y2/51
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µl
  • Pk:
    250 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: monoclonal
Clone: Y2/51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human