Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Catalog # ABCAAB8536-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:CD34 molecule
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:QBEND-10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Specifications
About this item
Mouse monoclonal [QBEND/10] to CD34.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEND/10
Conjugation:
Epitope: This antibody's target epitope is distinct from those of other anti-CD34 reactive mabs as demonstrated by its lack of sensitivity to neuraminidase treatment, and fluorescence staining profiles during flow cytometry analysis of target cell populations.
Host: Mouse
Isotype: IgG1
Reactivity: Human