Order Entry
Export
ContactUsLinkComponent
Anti-Nonspecific Cytotoxic Cells Mouse Monoclonal Antibody [clone: 5C.6]
Anti-Nonspecific Cytotoxic Cells Mouse Monoclonal Antibody [clone: 5C.6]
  ABCAAB2778-200
 :  Abcam
Anti-Nonspecific Cytotoxic Cells Mouse Monoclonal Antibody [clone: 5C.6]
  ABCAAB2778-200
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Nonspecific Cytotoxic Cells
  • Clonality:
    Monoclonal
  • Clone:
    5C.6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Western blot:
    Yes
  • Seq:
    1358
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µg
  • Pack type:
    Vial
  • Pk:
    200 µG

 

 

Mouse monoclonal [5C.6] to Nonspecific Cytotoxic Cells.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Nonspecific Cytotoxic Cells
Clonality: Monoclonal
Clone: 5C.6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Rat; Catfish; Bovine; Dog; Horse; Pig; Tilapia