Anti-Insulin Receptor beta Mouse Monoclonal Antibody [clone: 18-44]
ABCAAB983-50
: Abcam
- Antibody type:Primary
- Antigen name:Insulin Receptor beta
- Antigen symbol:INSRB
- Clonality:Monoclonal
- Clone:18-44
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Mouse monoclonal [18-44] to Insulin Receptor beta.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Insulin Receptor beta
Clonality: Monoclonal
Clone: 18-44
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human