Anti-HLA-MHC Class I Free Chain Mouse Monoclonal Antibody
Catalog # BIRBORB96190-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:HLA-MHC Class I Free Chain
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: HLA-MHC Class I Free Chain
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human