Order Entry
Export
ContactUsLinkComponent
Anti-HSP10 Rabbit Polyclonal Antibody
Anti-HSP10 Rabbit Polyclonal Antibody
Catalog # BIRBORB95894-50
Supplier:  Biorbyt
undefined
Anti-HSP10 Rabbit Polyclonal Antibody
Catalog # BIRBORB95894-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7,3, 0,02% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Immunochemistry: 10 ?g/ml: 1:1000

Type: Primary
Antigen: HSP10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: