Anti-BANF1 Rabbit Polyclonal Antibody
Catalog # BIRBORB87289-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:barrier to autointegration factor 1
- Antigen symbol:BANF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunofluorescence: 5 ?g/ml; Immunochemistry: 5 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: BANF1 (barrier to autointegration factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: