Anti-NOTCH1 Rabbit Polyclonal Antibody
100 µg
Catalog # BIRBORB77003-100
Supplier: Biorbyt
Jump to:
Product Details & Documents
ELISA: 1:1000-1:100000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: NOTCH1 (notch 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
orb77003
Datasheet
Specifications
- Size:100 µg
- Clonality:Polyclonal
- Purification:purified by affinity chromatography
- Antibody type:Primary
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Notch 1
- Cat. No.:BIRBORB77003-100
- Reactivity:Human|Rat|Mouse
- Immunogen:Peptide
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:NOTCH1
- ELISA:Yes



