Anti-Progesterone 11 alpha Mouse Monoclonal Antibody
BIRBORB76784-0.1
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Progesterone 11 alpha
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Storage buffer:Purified antibody containing PBS, 10mM sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Progesterone 11-alpha-BSA
- Purification:purified by Protein G chromatography
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Progesterone 11 alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: