Anti-GDP mannose 4,6 dehydratase Rabbit Polyclonal Antibody
Catalog # BIRBORB74125-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:GDP mannose 4,6 dehydratase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GDP mannose 4,6 dehydratase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human